Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Lieferant: Thermo Scientific
Synonyme:
Methyl 2-chloro-4-fluorophenylacetate, Methyl 2-(2-chloro-4-fluorophenyl)acetate
304780050
ACRO304780050EA
29.8
EUR
ACRO304780050
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Methyl-2-(2-chloro-4-fluorophenyl)acetate
Appearance: Clear colorless to light yellow Liquid
Formel:
C₉H₈ClFO₂ Molecular Weight: 202,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800611 CAS-Nummer: 214262-88-1 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5060 to 1.5080 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...