4'-Trifluormethoxyacetophenon 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-[4-(Trifluormethoxy)phenyl]-1-ethanon
306230050
ACRO306230050EA
86.9
EUR
ACRO306230050
4'-Trifluormethoxyacetophenon 98%
4'-Trifluormethoxyacetophenon
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol Siedepunkt: 110 °C (30 mmHg) Dichte: 1,278 g/cm³ Flammpunkt: 87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042404 CAS-Nummer: 85013-98-5 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...