Tris(4-fluorphenyl)phosphin 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
306340010
306340050
ACRO306340010EA
40.7
EUR
ACRO306340010
ACRO306340050
Tris(4-fluorphenyl)phosphin 98+%
Tris(4-fluorphenyl)phosphin
Formel:
C₁₈H₁₂F₃P Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013553 CAS-Nummer: 18437-78-0 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 79 to 83 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...