(Triphenylphosphoranylidene)acetaldehyde 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane

307490050 307490250 307491000
ACRO307490050EA 57.5 EUR
ACRO307490050 ACRO307490250 ACRO307491000
(Triphenylphosphoranylidene)acetaldehyde 97%
(Triphenylphosphoranylidene)acetaldehyde

CAS No.: 2136-75-6

Formel: C₂₀H₁₇OP
Molecular Weight: 304.33 g/mol
Schmelzpunkt: 185…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006994
CAS-Nummer: 2136-75-6

Bestellen

Spezifikation Testergebnisse

Appearance Cream to pink to orange to brown Crystalline powder
Infrared spectrum Conforms
Water ≤4.0 % (K.F.)
Titration with HClO4 96.0 to 104.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR