4-Aminostyrene 90% Stabilised, technical
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Vinylaniline
308480050
308480010
308480250
ACRO308480050EA
141
EUR
ACRO308480050
ACRO308480010
ACRO308480250
4-Aminostyrene 90% Stabilised, technical
4-Vinylanilin
Appearance: Yellow to orange-red Liquid or solid
Formel:
C₈H₉N Molecular Weight: 119.17 g/mol Siedepunkt: 249 °C (760 mmHg) Schmelzpunkt: 23…24 °C Dichte: 1.01 g/cm³ Flammpunkt: 99 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00015329 CAS-Nummer: 1520-21-4 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Yellow to orange-red Liquid or solid |
Infrared spectrum | Conforms |
GC | ≥88 % |
Stabilizer | Approx. 100 ppm 4-tert-Butylcatechol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...