4-Aminostyrene 90% Stabilised, technical

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Vinylaniline

308480050 308480010 308480250
ACRO308480050EA 141 EUR
ACRO308480050 ACRO308480010 ACRO308480250
4-Aminostyrene 90% Stabilised, technical
4-Vinylanilin

Appearance: Yellow to orange-red Liquid or solid

Formel: C₈H₉N
Molecular Weight: 119.17 g/mol
Siedepunkt: 249 °C (760 mmHg)
Schmelzpunkt: 23…24 °C
Dichte: 1.01 g/cm³
Flammpunkt: 99 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00015329
CAS-Nummer: 1520-21-4
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange-red Liquid or solid
Infrared spectrum Conforms
GC ≥88 %
Stabilizer Approx. 100 ppm 4-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR