2-Ethyl-1H-pyrrol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Ethyl-1H-pyrrole

308770010
ACRO308770010EA 92.9 EUR
ACRO308770010
2-Ethyl-1H-pyrrol 98%
2-Ethylpyrrole

Appearance: Clear yellow to brown Liquid

Formel: C₆H₉N
Molecular Weight: 95,15 g/mol
Siedepunkt: 142 °C (15 mmHg)
Dichte: 0,929 g/cm³
Flammpunkt: 62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060477
CAS-Nummer: 1551-06-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4985 to 1.5025 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR