Tetrafluorotetracyanoquinodimethane 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile
312670050
ACRO312670050EA
55.6
EUR
ACRO312670050
Tetrafluorotetracyanoquinodimethane 95%
Tetrafluorotetracyanoquinodimethane
Formel:
C₁₂F₄N₄ Molecular Weight: 276,15 g/mol Schmelzpunkt: 291 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042382 CAS-Nummer: 29261-33-4 UN: 3439 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow-green to orange-brown Powder |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...