Pentacarbonylrhenium(I)chlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

316310010 316310050
ACRO316310010EA 172 EUR
ACRO316310010 ACRO316310050
Pentacarbonylrhenium(I)chlorid 98%
Pentacarbonylrhenium(I)chlorid
Formel: C₅ClO₅Re
MDL: MFCD00013296
CAS-Nummer: 14099-01-5
EINECS: 237-948-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR