Triammoniumdodecamolybdophosphat Hydrat

Lieferant: Thermo Fisher Scientific
Achtung

317100500 317100010 317102500
ACRO317100500EA 71.8 EUR
ACRO317100500 ACRO317100010 ACRO317102500
Triammoniumdodecamolybdophosphat Hydrat
Triammoniumdodecamolybdophosphat Hydrat
Formel: H₁₂Mo₁₂N₃O₄₀P·H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00150423
CAS-Nummer: 54723-94-3
EINECS: 234-714-0
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤250 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR