Cadmiumacetat Dihydrat 98% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

317131000 317130025 317135000
ACRO317131000EA 49.3 EUR
ACRO317131000 ACRO317130025 ACRO317135000
Cadmiumacetat Dihydrat 98% zur Analyse
Cadmiumacetat Dihydrat
Formel: (H₃CCOO)₂Cd·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150020
CAS-Nummer: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 13,01614

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤10 ppm
Nitrate (NO3) ≤30 ppm
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm
Insoluble matter ≤0.005 % (in water)
Subst not pptd by H2S ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR