2-Amino-4-methyl-3-nitropyridin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyl-3-nitropyridin-2-amin, 4-Methyl-3-nitro-2-pyridylamin, 2-Amino-3-nitro-4-picolin

319550250
ACRO319550250EA 111 EUR
ACRO319550250
2-Amino-4-methyl-3-nitropyridin 98%
2-Amino-4-methyl-3-nitropyridin
Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 136…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006315
CAS-Nummer: 6635-86-5
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 141 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR