2-(Brommethyl)-2-(hydroxymethyl)-1,3-propandiol 99%

Lieferant: Thermo Scientific

Synonyme: 2-(Bromomethyl)-2-(hydroxymethyl)propane-1,3-diol

319630050
ACRO319630050EA 115 EUR
ACRO319630050
2-(Brommethyl)-2-(hydroxymethyl)-1,3-propandiol 99%
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol
Formel: C₅H₁₁BrO₃
Molecular Weight: 199,04 g/mol
Schmelzpunkt: 70…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135306
CAS-Nummer: 19184-65-7
EINECS: 242-863-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 77 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR