tert-Butylphosphonic acid 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-methylprop-2-ylphosphonic acid, Phosphonic acid, (1,1-dimethylethyl)-

321520010 321520050
ACRO321520010EA 51.8 EUR
ACRO321520010 ACRO321520050
tert-Butylphosphonic acid 98%
tert-Butylphosphonic acid

CAS No.: 4923-84-6

Formel: C₄H₁₁O₃P
Schmelzpunkt: 188…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012296
CAS-Nummer: 4923-84-6
UN: 3261
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 192 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR