2-[Ethyl(phenyl)amino]ethanol 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine, 2-(Ethyl(phenyl)amino)ethanol, N-Ethyl-N-2-ethanolaniline
325221000
ACRO325221000EA
94.2
EUR
ACRO325221000
2-[Ethyl(phenyl)amino]ethanol 96%
2-(N-Ethylanilino)ethanol
Formel:
C₁₀H₁₅NO Molecular Weight: 165,24 g/mol Siedepunkt: 276 °C Schmelzpunkt: 33 °C Flammpunkt: 120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020575 CAS-Nummer: 92-50-2 EINECS: 202-160-9 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid or solid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5640 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...