1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt

325560010 325560050
ACRO325560010EA 59.6 EUR
ACRO325560010 ACRO325560050
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt 95%
1,3,6,8-Pyrenetetrasulphonic acidtetrasodium salt

Appearance: Green Crystalline powder

Formel: C₁₆H₆Na₄O₁₂S₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00042029
CAS-Nummer: 59572-10-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to green Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR