3,4-Thiophendiamindihydrochlorid 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Thiophene-3,4-diamine dihydrochloride

325890010 325890050
ACRO325890010EA 70.5 EUR
ACRO325890010 ACRO325890050
3,4-Thiophendiamindihydrochlorid 96%
3,4-Thiophendiamindihydrochlorid

Appearance: Grey to brown Crystalline powder

Formel: C₄H₆N₂S
Schmelzpunkt: 240…245 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00671511
CAS-Nummer: 90069-81-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %
Loss on drying ≤3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR