3-Amino-N,N-dimethyl-4-nitroaniline 97%

Lieferant: Thermo Scientific
Achtung

326220050
ACRO326220050EA 96.2 EUR
ACRO326220050
3-Amino-N,N-dimethyl-4-nitroaniline 97%
3-Amino-N,N-dimethyl-4-nitroaniline

Appearance: Yellow-brown Powder

Formel: C₈H₁₁N₃O₂
Schmelzpunkt: 134…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320677
CAS-Nummer: 2069-71-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-brown Powder
Infrared spectrum Conforms
Melting point 134 to 139 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR