4-Brom-3-nitroanilin 97%

Lieferant: Thermo Scientific
Achtung

326240050
ACRO326240050EA 91.5 EUR
ACRO326240050
4-Brom-3-nitroanilin 97%
4-Brom-3-nitroanilin
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
Schmelzpunkt: 128…132 °C
MDL: MFCD01320678
CAS-Nummer: 53324-38-2

Bestellen

Spezifikation Testergebnisse

Appearance Orange to dark brown Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR