Wasser, ECD tested for pesticide analysis Residual

Lieferant: Thermo Fisher Scientific

Synonyme: Wasserstoffoxid, Hydrogenoxid

326650010 326650025
ACRO326650010EA 32.3 EUR
ACRO326650010 ACRO326650025
Wasser, ECD tested for pesticide analysis Residual
Wasser
Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Additional info residue (at bottling) : < 0.0005%
Color scale ≤10 APHA
GC/ECD (as lindane) ≤5 ng/l
Residue after evaporation ≤0.0002 %
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR