X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid) 99+% für die Molekularbiologie DNAse-, RNAse-, proteasefrei
Lieferant: Thermo Fisher Scientific
Synonyme:
BCIG
327241000
327245000
ACRO327241000EA
65.3
EUR
ACRO327241000
ACRO327245000
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid) 99+% für die Molekularbiologie DNAse-, RNAse-, proteasefrei
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel:
C₁₄H₁₅BrClNO₆ Molecular Weight: 408.63 g/mol Schmelzpunkt: 230 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005666 CAS-Nummer: 7240-90-6 EINECS: 230-640-8 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤0.2 % |
TLC | ≥99.0 % |
Specific optical rotation | -61° to -63° (20°C, 589 nm) (c=1, H2O/DMF 1/1) |
Dnase,Rnase,Protease activity | none detected |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...