Tris EDTA buffer, for molecular biology, DNAse, RNAse, protease free ready to use, pH 8,0
Lieferant: Thermo Fisher Scientific
327345000
ACRO327345000EA
98.5
EUR
ACRO327345000
Tris EDTA buffer, for molecular biology, DNAse, RNAse, protease free ready to use, pH 8,0
Puffer
Biologische Puffer
CAS No.: 38641-82-6
MDL No.: MFCD00236359
Appearance: Clear colourless solution
Storage temperature: Ambient
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...