CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

327741000 327740010 327740250
ACRO327741000EA 97.7 EUR
ACRO327741000 ACRO327740010 ACRO327740250
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) 99%
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel: C₉H₁₉NO₄S
Molecular Weight: 237.32 g/mol
Schmelzpunkt: 263…271 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041778
CAS-Nummer: 73463-39-5

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Solubility (0.1M in water) Clear colorless solution
UV at 280 nm A: ≤0.03
pH 5 to 6.5 (1 % in water at 25°C)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR