Fumonisin B2 96+%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,2,3-Propanetricarboxylic acid, 1,1?-[1-(12-amino-4,11-dihydroxy-2-methyl-tridecyl)-2-(1-methylpentyl)-1,2-ethane-diyl]ester, Fumonisin B2 from Fusarium moniliforme, sphingosine N-acyltransferase inhibitor, 1,2,3-Propanetricarboxylic acid, 1,1'-(1-(2-amino-9,11-dihydroxy-2-methyltridecyl)-2-(1-methylpentyl)-1,2-ethanediyl) ester

328260010
ACRO328260010EA 416 EUR
ACRO328260010
Fumonisin B2 96+%
Fumonisin B2
Formel: C₃₄H₅₉NO₁₄
Molecular Weight: 705,839 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00133350
CAS-Nummer: 116355-84-1
UN: 2811
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to brown Solid
HPLC ≥96.0 %
Elemental Analysis ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR