Rottlerin 95+%

Lieferant: Thermo Scientific
Achtung

Synonyme: (E)-1-(6-(3-Acetyl-2,4,6-trihydroxy-5-methylbenzyl)-5,7-dihydroxy-2,2-dimethyl-2H-chromen-8-yl)-3-phenylprop-2-en-1-one

328490100
ACRO328490100EA 98.3 EUR
ACRO328490100
Rottlerin 95+%
Rottlerin

Appearance: Orange to brown Powder

Formel: C₃₀H₂₈O₈
Molecular Weight: 516,546 g/mol
Schmelzpunkt: >200 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00017361
CAS-Nummer: 82-08-6

Bestellen

Spezifikation Testergebnisse

Appearance Orange to brown Powder
Infrared spectrum Conforms
HPLC ≥95.0 %
TLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR