Staurosporine 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one, Antibiotic AM-2282

328532500 328530010
ACRO328532500EA 298 EUR
ACRO328532500 ACRO328530010
Staurosporine 98%
Staurosporine

CAS No.: 62996-74-1

Formel: C₂₈H₂₆N₄O₃
Molecular Weight: 466.541 g/mol
Schmelzpunkt: 270 °C
Lagertemperatur: Kühlschrank
MDL: MFCD18252446
CAS-Nummer: 62996-74-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige Powder
HPLC =97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR