Griess Reagenz

Lieferant: Thermo Scientific
Achtung

328670500
ACRO328670500EA 77.2 EUR
ACRO328670500
Griess Reagenz
Griess Reagenz

Appearance: Clear colorless to light pink Solution

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light pink Solution
Suitability (for NO2 det.in biol.media) Passes test
Composition in Phosphoric acid (5%)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR