(±)-Ibotenic acid monohydrate 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (±)-alpha-Amino-3-hydroxy-5-isoxazoleacetic acid monohydrate

329130010
ACRO329130010EA 316 EUR
ACRO329130010
(±)-Ibotenic acid monohydrate 95%
(±)-Ibotenic acid monohydrate

Appearance: White to off-white Solid

Formel: C₅H₈N₂O₅
Molecular Weight: 176,128 g/mol
Schmelzpunkt: 141…147 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00149579
CAS-Nummer: 60573-88-8
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
HPLC ≥94 %
NMR Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR