N-Hydroxyphthalimid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Hydroxyphthalsäureimid, N-Hydroxy-1,2-benzoldicarbonsäureimid

329871000 329875000 329870025
ACRO329871000EA 35.9 EUR
ACRO329871000 ACRO329875000 ACRO329870025
N-Hydroxyphthalimid 98%
N-Hydroxyphthalimid
Formel: C₈H₅NO₃
Molecular Weight: 163.13 g/mol
Siedepunkt: 412
Schmelzpunkt: 233…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005891
CAS-Nummer: 524-38-9
EINECS: 208-358-1
Merck Index: 13,04864

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Water ≤2 %
Titration with NaOH 97.5 to 102.5 % (On anhydrous substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR