1-Boc-4-piperidon 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

330270050 330270250 330271000
ACRO330270050EA 56 EUR
ACRO330270050 ACRO330270250 ACRO330271000
1-Boc-4-piperidon 99%
1-Boc-4-piperidon
Formel: C₁₀H₁₇NO₃
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151800
CAS-Nummer: 79099-07-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR