5-Fluor-2-indolcarbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Fluoro-1H-indole-2-carboxylic acid

331590010 331590050
ACRO331590010EA 44.3 EUR
ACRO331590010 ACRO331590050
5-Fluor-2-indolcarbonsäure 98%
5-Fluoroindole-2-carboxylic acid
Formel: C₉H₆FNO₂
Schmelzpunkt: 254…258 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 254 to 258 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR