Ethyl (S)-(-)-2-methoxypropionate 98%

Lieferant: Thermo Scientific

331730100
ACRO331730100EA 45.3 EUR
ACRO331730100
Ethyl (S)-(-)-2-methoxypropionate 98%
Ethyl (s)-(-)-2-methoxypropionate

Appearance: Clear colorless to light yellow Liquid

Formel: C₆H₁₂O₃
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632590
CAS-Nummer: 41918-08-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -86° to -92° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR