(R)-2-Methoxypropanoic acid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

331810010
ACRO331810010EA 222 EUR
ACRO331810010
(R)-2-Methoxypropanoic acid 97%
(R)-2-Methoxypropanoic acid

Appearance: Clear colorless Liquid

Formel: C₄H₈O₃
Dichte: 1,094 g/cm³
Flammpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632582
CAS-Nummer: 23943-96-6
UN: 3265
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4132 to 1.4152 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +76° to +80° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR