4-Chlor-o-phenylendiamin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
331860050
331860250
331861000
ACRO331860050EA
30.2
EUR
ACRO331860050
ACRO331860250
ACRO331861000
4-Chlor-o-phenylendiamin 97%
4-Chlor-o-phenylendiamin
Formel:
C₆H₇ClN₂ Siedepunkt: 280 °C (1013 hPa) Schmelzpunkt: 70…75 °C |
MDL:
MFCD00011691 CAS-Nummer: 95-83-0 EINECS: 202-456-8 |
Spezifikation Testergebnisse
Appearance | Grey-brown to dark brown Powder or granules and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 75 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...