Diethyl (4-nitrobenzyl)phosphonate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(4-Nitrobenzyl)phosphonic acid diethyl ester, Diethyl 4-nitrobenzylphosphonate
332290050
332290250
ACRO332290050EA
91.9
EUR
ACRO332290050
ACRO332290250
Diethyl (4-nitrobenzyl)phosphonate 98%
Diethyl (4-nitrobenzyl)phosphonate
Appearance: Clear yellow to orange to dark red Liquid
Formel:
C₁₁H₁₆NO₅P Siedepunkt: 151 °C (,1 mmHg) Dichte: 1,24 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047747 CAS-Nummer: 2609-49-6 |
Spezifikation Testergebnisse
Appearance | Clear yellow to orange to dark red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5220 to 1.5260 (20°C, 589 nm) |
NMR | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...