Carboxymethylcellulose Natriumsalz, M.W. ∼ 700000 (DS=0.9)

Lieferant: Thermo Fisher Scientific

Synonyme: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

332640010 332641000
ACRO332640010EA 133 EUR
ACRO332640010 ACRO332641000
Carboxymethylcellulose Natriumsalz, M.W. ∼ 700000 (DS=0.9)
Carboxymethyl cellulose sodium salt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081472
CAS-Nummer: 9004-32-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Assay ≥99.5 %
Loss on drying ≤10 % (As packed) (3 to 5 g, 105°C, 2 h)
Heavy metals ≤20 ppm
Additional info degree of substitution: 0.80 to 0.95
Degree of substitution 0.80 to 0.95
Viscosity 2500 to 4500 mPa.s (1 % at 25°C) (Brookfield)
pH 6.5 to 8 (1 % solution)
Sodium chloride (NaCl) ≤0.25 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Impurity ≤0.4 % (Sodium glycolate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR