2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Mercapto-2-methyltetrahydrofuran, 2-Methyl-3-tetrahydrofuranthiol
332700250
ACRO332700250EA
242
EUR
ACRO332700250
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 97%
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture)
Appearance: Clear colorless to light yellow Liquid
Formel:
C₅H₁₀OS Molecular Weight: 118,19 g/mol Siedepunkt: 160…180 °C (760 mmHg) Dichte: 1,04 g/cm³ Flammpunkt: 40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632596 CAS-Nummer: 57124-87-5 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4730 to 1.4910 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...