(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 99%

Lieferant: Thermo Fisher Scientific
Achtung

333200010 333200050
ACRO333200010EA 51.7 EUR
ACRO333200010 ACRO333200050
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 99%
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid

Appearance: White Crystalline powder or crystals

Formel: C₁₃H₁₈O₂
Schmelzpunkt: 49…53 °C
Flammpunkt: 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00069289
CAS-Nummer: 51146-56-6

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥98.5 %
Specific optical rotation +57° to +61° (20 °C, 589 nm) (c=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR