N-Boc-DL-2-aminotetralin-2-carboxylic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Boc-amino)-1,2,3,4-tetrahydronaphthalene-2-carboxylic Acid, 2-N-Boc-amino-tetrahydro-2-naphthoic acid

334370010
ACRO334370010EA 134 EUR
ACRO334370010
N-Boc-DL-2-aminotetralin-2-carboxylic acid 98%
N-Boc-DL-2-aminotetralin-2-carboxylic acid

Appearance: White to beige Crystalline powder

Formel: C₁₆H₂₁NO₄
Molecular Weight: 291,347 g/mol
Schmelzpunkt: 177…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800586
CAS-Nummer: 98569-12-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR