Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride 98%

Lieferant: Thermo Fisher Scientific

334410010
ACRO334410010EA 244 EUR
ACRO334410010
Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride 98%
Ethyl-trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride

Appearance: White to beige to grey Powder

Formel: C₉H₁₆ClNO₂
Molecular Weight: 205.682 g/mol
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863252
CAS-Nummer: 142547-16-8

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to grey Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR