2-Brom-5-nitroanilin 98%

Lieferant: Thermo Fisher Scientific
Achtung

334860050 334860250
ACRO334860050EA 51.3 EUR
ACRO334860050 ACRO334860250
2-Brom-5-nitroanilin 98%
2-Brom-5-nitroanilin
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217.02 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051578
CAS-Nummer: 10403-47-1
EINECS: 233-874-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-green to yellow-brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 137 to 142 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR