3-Amino-4,4-dimethylpentanoic acid hydrate 98%
Lieferant: Thermo Scientific
335855000
ACRO335855000EA
138
EUR
ACRO335855000
3-Amino-4,4-dimethylpentanoic acid hydrate 98%
3-Amino-4,4-dimethylpentanoic acid hydrate
Appearance: White Crystalline flakes
Formel:
C₇H₁₇NO₃ Schmelzpunkt: 223…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791052 CAS-Nummer: 336185-29-6 |
Spezifikation Testergebnisse
Appearance | White Crystalline flakes |
Infrared spectrum | Conforms |
Water | ≤14 % (K.F.) |
Titration with HClO4 | ≥97.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...