Diethylallylphosphonat 97%

Lieferant: Thermo Fisher Scientific

336110050 336110250
ACRO336110050EA 80.3 EUR
ACRO336110050 ACRO336110250
Diethylallylphosphonat 97%
Diethylallylphosphonat
Formel: C₇H₁₅O₃P
Siedepunkt: 97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015134
CAS-Nummer: 1067-87-4

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4325 to 1.4345 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR