4-Methylumbelliferyl-β-D-xylopyranoside 99%
Lieferant: Thermo Scientific
Synonyme:
4-Methyl-7-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, 4-Methylumbelliferyl-β-D-xylopyranoside, β-xylosidase substrate
337272500
ACRO337272500EA
256
EUR
ACRO337272500
4-Methylumbelliferyl-β-D-xylopyranoside 99%
4-Methylumbelliferyl-beta-D-xylopyranoside
Appearance: White to almost white Powder
Formel:
C₁₅H₁₆O₇ Molecular Weight: 308,286 g/mol Schmelzpunkt: 223 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00037607 CAS-Nummer: 6734-33-4 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
HPLC | ≥98.5 % |
Appearance of solution | (1% in DMF) Almost clear, colorless to light yellow |
Specific optical rotation | -28° to -32° (20°C, 589 nm) (c=0.3, pyridine) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...