Dimethyl-(2-hydroxyethyl)phosphonat  95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-Hydroxyethyl)phosphonic Acid Dimethyl Ester

338160050 338160250
ACRO338160050EA 102 EUR
ACRO338160050 ACRO338160250
Dimethyl-(2-hydroxyethyl)phosphonat  95%
Dimethyl(2-hydroxyethyl)phosphonate

Appearance: Clear colorless to light yellow Liquid

Formel: C₄H₁₁O₄P
Siedepunkt: 98…104 °C (,1 mmHg)
Dichte: 1,247 g/cm³
Flammpunkt: 192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043376
CAS-Nummer: 54731-72-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4460 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR