Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride 60% in Methanol

Lieferant: Thermo Scientific
Gefahr

338531000
ACRO338531000EA 101 EUR
ACRO338531000
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride 60% in Methanol
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride

Appearance: Clear yellow Solution

Formel: C₂₆H₅₈ClNO₃Si
Molecular Weight: 496,289 g/mol
Dichte: 0,89 g/cm³
Flammpunkt: 15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00054224
CAS-Nummer: 27668-52-6
UN: 1230
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Solution
Infrared spectrum Conforms
Titration Argentometric ≥58 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR