Trimethyl(vinyloxy)silane 97%

Lieferant: Thermo Fisher Scientific
Gefahr

338700010 338700050
ACRO338700010EA 30.4 EUR
ACRO338700010 ACRO338700050
Trimethyl(vinyloxy)silane 97%
Trimethyl(vinyloxy)silane

Appearance: Clear light yellow Liquid

Formel: C₅H₁₂OSi
Siedepunkt: 74…75 °C (760 mmHg)
Dichte: 0,77 g/cm³
Flammpunkt: –10 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00054764
CAS-Nummer: 6213-94-1
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR