Trimethyl(vinyloxy)silane 97%
Lieferant: Thermo Fisher Scientific
338700010
338700050
ACRO338700010EA
30.4
EUR
ACRO338700010
ACRO338700050
Trimethyl(vinyloxy)silane 97%
Appearance: Clear light yellow Liquid
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3885 to 1.3905 (20°C, 589 nm) |
GC | ≥96.0 % |