3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-3-(2,2-Dimethyl-1,3-dioxolan-4-yl)propan-1-ol
339610010
ACRO339610010EA
90.3
EUR
ACRO339610010
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 97%
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol
Appearance: Clear colorless to yellow Viscous liquid
Formel:
C₈H₁₆O₃ Molecular Weight: 160,213 g/mol Siedepunkt: 90 °C (10 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631673 CAS-Nummer: 51268-87-2 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4410 to 1.4430 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific optical rotation | +9° to +12° (20°C, 589 nm) (c=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...