Formononetin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 7-Hydroxy-3-(4-methoxyphenyl)chromone, 7-Hydroxy-4'-methoxyisoflavone

342480010
ACRO342480010EA 106 EUR
ACRO342480010
Formononetin 98%
Formononetin

Appearance: White to light beige Powder

Formel: C₁₆H₁₂O₄
Schmelzpunkt: 256…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016948
CAS-Nummer: 485-72-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 256 to 261 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR